Learn More
3-(4-Bromophenyl)propionic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2678506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(4-Bromophenyl)propionic acid | |
133°C to 136°C | |
MFCD01310793 | |
3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm | |
C1=CC(=CC=C1CCC(=O)O)Br | |
229.073 | |
229.07 |
1643-30-7 | |
C9H9BrO2 | |
5 g | |
NCSTWHYWOVZDOC-UHFFFAOYSA-N | |
3-(4-bromophenyl)propanoic acid | |
2735609 | |
98% |
Chemical Identifiers
1643-30-7 | |
229.073 | |
NCSTWHYWOVZDOC-UHFFFAOYSA-N | |
2735609 | |
C1=CC(=CC=C1CCC(=O)O)Br |
C9H9BrO2 | |
MFCD01310793 | |
3-4-bromophenyl propionic acid, 3-4-bromophenyl propanoic acid, benzenepropanoic acid, 4-bromo, 3-4-bromo-phenyl-propionic acid, p-bromohydrocinnamic acid, 3-4-bromophenyl-propionic acid, 3-4-bromophenyl propanoicacid, pubchem19769, 4-bromohydrocinnamic acid, acmc-1c0vm | |
3-(4-bromophenyl)propanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only