missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-9-phenylcarbazole, 98%

$29.45 - $345.43

Chemical Identifiers

CAS 1028647-93-9
Molecular Formula C24H16BrN
Molecular Weight (g/mol) 398.303
MDL Number MFCD14582939
InChI Key JEYLGFCAZBGCMC-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
PubChem CID 51358293
IUPAC Name 3-(4-bromophenyl)-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64542MD View Documents Alfa Aesar™
H64542MD
250mg Each for $29.45
 
AAH6454203 View Documents Alfa Aesar™
H6454203
1g Each for $86.99
 
AAH6454206 View Documents Alfa Aesar™
H6454206
5g Each for $345.43
 
Specifications

Chemical Identifiers

1028647-93-9
398.303
JEYLGFCAZBGCMC-UHFFFAOYSA-N
51358293
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
C24H16BrN
MFCD14582939
3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
3-(4-bromophenyl)-9-phenylcarbazole

Specifications

250mg
C24H16BrN
19325695
JEYLGFCAZBGCMC-UHFFFAOYSA-N
3-(4-bromophenyl)-9-phenylcarbazole
51358293
Solid
3-(4-Bromophenyl)-9-phenylcarbazole
1028647-93-9
MFCD14582939
3-4-bromophenyl-9-phenyl-9h-carbazole, 3-4-bromophenyl-9-phenylcarbazole, 3-4-bromophenyl-n-phenylcarbazole, 3-4-bromophenyl-9-phenyl9h-carbazole, pubchem23233, 3-4-bromophenyl-9-phenyl-carbazole, 3-4-bromo-phenyl-9-phenyl-9h-carbazole
C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC=C(C=C4)Br)C5=CC=CC=C52
398.303
398.3
98%