missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1858241-81-2
Molecular Formula C15H12BrN3
Molecular Weight (g/mol) 314.186
MDL Number MFCD22683118
InChI Key TYXCTRHZBUPEHB-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
PubChem CID 98000572
IUPAC Name 3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60327MC View Documents Alfa Aesar™
H60327MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60327ME View Documents Alfa Aesar™
H60327ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1858241-81-2
314.186
TYXCTRHZBUPEHB-UHFFFAOYSA-N
98000572
CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br
C15H12BrN3
MFCD22683118
3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole

Specifications

100mg
C15H12BrN3
3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br
314.186
314.18
3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole
1858241-81-2
MFCD22683118
TYXCTRHZBUPEHB-UHFFFAOYSA-N
3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
98000572
97%