missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-5-methyl-4-(3-nitrophenyl)-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1421263-13-9
Molecular Formula C15H11BrN4O2
Molecular Weight (g/mol) 359.183
MDL Number MFCD22683126
InChI Key WQZPCZDVKCPFRR-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-5-methyl-4-3-nitrophenyl-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-3-nitrophenyl-4h-1,2,4-triazole
PubChem CID 97290228
IUPAC Name 3-(4-bromophenyl)-5-methyl-4-(3-nitrophenyl)-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC(=CC=C2)[N+](=O)[O-])C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60457MC View Documents Alfa Aesar™
H60457MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60457ME View Documents Alfa Aesar™
H60457ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421263-13-9
359.183
WQZPCZDVKCPFRR-UHFFFAOYSA-N
97290228
CC1=NN=C(N1C2=CC(=CC=C2)[N+](=O)[O-])C3=CC=C(C=C3)Br
C15H11BrN4O2
MFCD22683126
3-4-bromophenyl-5-methyl-4-3-nitrophenyl-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-3-nitrophenyl-4h-1,2,4-triazole
3-(4-bromophenyl)-5-methyl-4-(3-nitrophenyl)-1,2,4-triazole

Specifications

100mg
C15H11BrN4O2
3-4-bromophenyl-5-methyl-4-3-nitrophenyl-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-3-nitrophenyl-4h-1,2,4-triazole
CC1=NN=C(N1C2=CC(=CC=C2)[N+](=O)[O-])C3=CC=C(C=C3)Br
359.183
359.18
3-(4-Bromophenyl)-5-methyl-4-(3-nitrophenyl)-4H-1,2,4-triazole
1421263-13-9
MFCD22683126
WQZPCZDVKCPFRR-UHFFFAOYSA-N
3-(4-bromophenyl)-5-methyl-4-(3-nitrophenyl)-1,2,4-triazole
97290228
97%