missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Bromophenyl)-5-methyl-1,2,4-oxadiazole, 97%, Maybridge

Chemical Identifiers

CAS 118183-92-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H7BrN2O
Formula Weight 239.07
MDL Number MFCD00487341
Chemical Name or Material 3-(4-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC34810DA SDS Fisher Scientific Company
CC34810DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34810DE Fisher Scientific Company
CC34810DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

118183-92-9
100.0
C9H7BrN2O
MFCD00487341
97%
96.5
97%
239.07
3-(4-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole

Specifications

118183-92-9
100.0
1g
C9H7BrN2O
MFCD00487341
97%
96.5
Amber glass bottle
97%
239.07
3-(4-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.