missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(4-methoxybenzyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1858241-84-5
Molecular Formula C17H16BrN3O
Molecular Weight (g/mol) 358.239
MDL Number MFCD22683153
InChI Key YMJHVVWEYLBWNG-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-4-methoxyphenyl methyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-methoxybenzyl-5-methyl-4h-1,2,4-triazole
PubChem CID 98000593
IUPAC Name 3-(4-bromophenyl)-4-[(4-methoxyphenyl)methyl]-5-methyl-1,2,4-triazole
SMILES CC1=NN=C(N1CC2=CC=C(C=C2)OC)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60918MC View Documents Alfa Aesar™
H60918MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60918ME View Documents Alfa Aesar™
H60918ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1858241-84-5
358.239
YMJHVVWEYLBWNG-UHFFFAOYSA-N
98000593
CC1=NN=C(N1CC2=CC=C(C=C2)OC)C3=CC=C(C=C3)Br
C17H16BrN3O
MFCD22683153
3-4-bromophenyl-4-4-methoxyphenyl methyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-methoxybenzyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-[(4-methoxyphenyl)methyl]-5-methyl-1,2,4-triazole

Specifications

100mg
C17H16BrN3O
3-4-bromophenyl-4-4-methoxyphenyl methyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-methoxybenzyl-5-methyl-4h-1,2,4-triazole
CC1=NN=C(N1CC2=CC=C(C=C2)OC)C3=CC=C(C=C3)Br
358.239
358.24
3-(4-Bromophenyl)-4-(4-methoxybenzyl)-5-methyl-4H-1,2,4-triazole
1858241-84-5
MFCD22683153
YMJHVVWEYLBWNG-UHFFFAOYSA-N
3-(4-bromophenyl)-4-[(4-methoxyphenyl)methyl]-5-methyl-1,2,4-triazole
98000593
97%