missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(4-ethylphenyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1421263-53-7
Molecular Formula C17H16BrN3
Molecular Weight (g/mol) 342.24
MDL Number MFCD22683119
InChI Key FSFAVMUCBYVKSQ-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-4h-1,2,4-triazole
PubChem CID 97030404
IUPAC Name 3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole
SMILES CCC1=CC=C(C=C1)N2C(=NN=C2C3=CC=C(C=C3)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60339MC View Documents Alfa Aesar™
H60339MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60339ME View Documents Alfa Aesar™
H60339ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421263-53-7
342.24
FSFAVMUCBYVKSQ-UHFFFAOYSA-N
97030404
CCC1=CC=C(C=C1)N2C(=NN=C2C3=CC=C(C=C3)Br)C
C17H16BrN3
MFCD22683119
3-4-bromophenyl-4-4-ethylphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole

Specifications

100mg
C17H16BrN3
3-4-bromophenyl-4-4-ethylphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-ethylphenyl-5-methyl-4h-1,2,4-triazole
CCC1=CC=C(C=C1)N2C(=NN=C2C3=CC=C(C=C3)Br)C
342.24
342.24
3-(4-Bromophenyl)-4-(4-ethylphenyl)-5-methyl-4H-1,2,4-triazole
1421263-53-7
MFCD22683119
FSFAVMUCBYVKSQ-UHFFFAOYSA-N
3-(4-bromophenyl)-4-(4-ethylphenyl)-5-methyl-1,2,4-triazole
97030404
97%