missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(4-chlorophenyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1421261-95-1
Molecular Formula C15H11BrClN3
Molecular Weight (g/mol) 348.628
MDL Number MFCD22683116
InChI Key VHRRICGRDAJVCU-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-4-chlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-chlorophenyl-5-methyl-4h-1,2,4-triazole
PubChem CID 97030402
IUPAC Name 3-(4-bromophenyl)-4-(4-chlorophenyl)-5-methyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60295MC View Documents Alfa Aesar™
H60295MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60295ME View Documents Alfa Aesar™
H60295ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421261-95-1
348.628
VHRRICGRDAJVCU-UHFFFAOYSA-N
97030402
CC1=NN=C(N1C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Br
C15H11BrClN3
MFCD22683116
3-4-bromophenyl-4-4-chlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-chlorophenyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-(4-chlorophenyl)-5-methyl-1,2,4-triazole

Specifications

100mg
C15H11BrClN3
3-4-bromophenyl-4-4-chlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-4-chlorophenyl-5-methyl-4h-1,2,4-triazole
CC1=NN=C(N1C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Br
348.628
348.63
3-(4-Bromophenyl)-4-(4-chlorophenyl)-5-methyl-4H-1,2,4-triazole
1421261-95-1
MFCD22683116
VHRRICGRDAJVCU-UHFFFAOYSA-N
3-(4-bromophenyl)-4-(4-chlorophenyl)-5-methyl-1,2,4-triazole
97030402
97%