missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1858255-54-5
Molecular Formula C17H16BrN3O2
Molecular Weight (g/mol) 374.238
MDL Number MFCD22683127
InChI Key SPFFEOOTWDLJSH-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole
PubChem CID 98000573
IUPAC Name 3-(4-bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60477MC View Documents Alfa Aesar™
H60477MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60477ME View Documents Alfa Aesar™
H60477ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1858255-54-5
374.238
SPFFEOOTWDLJSH-UHFFFAOYSA-N
98000573
CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br
C17H16BrN3O2
MFCD22683127
3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-1,2,4-triazole

Specifications

100mg
C17H16BrN3O2
3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole
CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br
374.238
374.24
3-(4-Bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-4H-1,2,4-triazole
1858255-54-5
MFCD22683127
SPFFEOOTWDLJSH-UHFFFAOYSA-N
3-(4-bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-1,2,4-triazole
98000573
97%