missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1421261-47-3
Molecular Formula C15H10BrCl2N3
Molecular Weight (g/mol) 383.07
MDL Number MFCD22683144
InChI Key YHMKSEQFQPFOLL-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-4h-1,2,4-triazole
PubChem CID 97030413
IUPAC Name 3-(4-bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60713MC View Documents Alfa Aesar™
H60713MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60713ME View Documents Alfa Aesar™
H60713ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421261-47-3
383.07
YHMKSEQFQPFOLL-UHFFFAOYSA-N
97030413
CC1=NN=C(N1C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)Br
C15H10BrCl2N3
MFCD22683144
3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-1,2,4-triazole

Specifications

100mg
C15H10BrCl2N3
3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dichlorophenyl-5-methyl-4h-1,2,4-triazole
CC1=NN=C(N1C2=CC(=C(C=C2)Cl)Cl)C3=CC=C(C=C3)Br
383.07
383.07
3-(4-Bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-4H-1,2,4-triazole
1421261-47-3
MFCD22683144
YHMKSEQFQPFOLL-UHFFFAOYSA-N
3-(4-bromophenyl)-4-(3,4-dichlorophenyl)-5-methyl-1,2,4-triazole
97030413
97%