missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromophenyl)-4-(2-methoxyphenyl)-5-methyl-4H-1,2,4-triazole, 97%

Chemical Identifiers

CAS 1421261-61-1
Molecular Formula C16H14BrN3O
Molecular Weight (g/mol) 344.212
MDL Number MFCD22683149
InChI Key CCXAVTXHPGUSRI-UHFFFAOYSA-N
Synonym 3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-4h-1,2,4-triazole
PubChem CID 97030415
IUPAC Name 3-(4-bromophenyl)-4-(2-methoxyphenyl)-5-methyl-1,2,4-triazole
SMILES CC1=NN=C(N1C2=CC=CC=C2OC)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60836MC View Documents Alfa Aesar™
H60836MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH60836ME View Documents Alfa Aesar™
H60836ME
500mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421261-61-1
344.212
CCXAVTXHPGUSRI-UHFFFAOYSA-N
97030415
CC1=NN=C(N1C2=CC=CC=C2OC)C3=CC=C(C=C3)Br
C16H14BrN3O
MFCD22683149
3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-4h-1,2,4-triazole
3-(4-bromophenyl)-4-(2-methoxyphenyl)-5-methyl-1,2,4-triazole

Specifications

100mg
C16H14BrN3O
3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-2-methoxyphenyl-5-methyl-4h-1,2,4-triazole
CC1=NN=C(N1C2=CC=CC=C2OC)C3=CC=C(C=C3)Br
344.212
344.21
3-(4-Bromophenyl)-4-(2-methoxyphenyl)-5-methyl-4H-1,2,4-triazole
1421261-61-1
MFCD22683149
CCXAVTXHPGUSRI-UHFFFAOYSA-N
3-(4-bromophenyl)-4-(2-methoxyphenyl)-5-methyl-1,2,4-triazole
97030415
97%