missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC59510DE

 View more versions of this product

Catalog No. CC59510DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-(4-Bromophenoxy)-N, N-dimethylpropylamine
96.5
97%
C11H16BrNO
5g
CAPNMXRLKVMOOB-UHFFFAOYSA-N
3-(4-bromophenoxy)-N,N-dimethylpropan-1-amine
2183674
97%
76579-64-1
100.0
Amber Glass Bottle
MFCD03718813
3-4-bromophenoxy propyl dimethylamine, 3-4-bromophenoxy-n,n-dimethylpropylamine, 3-4-bromophenoxy-n,n-dimethylpropan-1-amine, 1-propanamine,3-4-bromophenoxy-n,n-dimethyl, 4-n,n-dimethylaminopropyloxy phenyl bromide, n,n-dimethyl-3-4-bromophenoxy-1-propanamine
CN(C)CCCOC1=CC=C(C=C1)Br
258.159
258.16

Chemical Identifiers

76579-64-1
258.159
CAPNMXRLKVMOOB-UHFFFAOYSA-N
2183674
CN(C)CCCOC1=CC=C(C=C1)Br
C11H16BrNO
MFCD03718813
3-4-bromophenoxy propyl dimethylamine, 3-4-bromophenoxy-n,n-dimethylpropylamine, 3-4-bromophenoxy-n,n-dimethylpropan-1-amine, 1-propanamine,3-4-bromophenoxy-n,n-dimethyl, 4-n,n-dimethylaminopropyloxy phenyl bromide, n,n-dimethyl-3-4-bromophenoxy-1-propanamine
3-(4-bromophenoxy)-N,N-dimethylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions