missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Bromobenzoyl)propionic Acid, TCI America™

$93.20 - $322.38

Chemical Identifiers

CAS 6340-79-0
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.083
MDL Number MFCD00016563
InChI Key ZODFRCZNTXLDDW-UHFFFAOYSA-N
Synonym 3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
PubChem CID 80646
IUPAC Name 4-(4-bromophenyl)-4-oxobutanoic acid
SMILES C1=CC(=CC=C1C(=O)CCC(=O)O)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31295G View Documents TCI America
B31295G
5g Each for $93.20
 
B312925G View Documents TCI America
B312925G
25g Each for $322.38
 
Specifications

Chemical Identifiers

6340-79-0
257.083
ZODFRCZNTXLDDW-UHFFFAOYSA-N
80646
C1=CC(=CC=C1C(=O)CCC(=O)O)Br
C10H9BrO3
MFCD00016563
3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
4-(4-bromophenyl)-4-oxobutanoic acid

Specifications

Undesignated
5g
C10H9BrO3
3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
C1=CC(=CC=C1C(=O)CCC(=O)O)Br
257.083
257.08
≥98.0% (HPLC,T)
150°C
6340-79-0
MFCD00016563
ZODFRCZNTXLDDW-UHFFFAOYSA-N
4-(4-bromophenyl)-4-oxobutanoic acid
80646
Crystal-Powder at 20°C
3-(4-Bromobenzoyl)propionic Acid
Safety and Handling

Safety and Handling

TSCA : No