Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Bromobenzoyl)propionic acid, 98+%

$43.55 - $378.66

Chemical Identifiers

Chemical Name or Material 3-(4-Bromobenzoyl)propionic acid
CAS 6340-79-0
Molecular Formula C10H9BrO3
Formula Weight 257.09
MDL Number MFCD00016563
Beilstein 2104410
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0400803

 
alfa aesar™
L0400803
1g Each for $43.55

AAL0400814

 
alfa aesar™
L0400814
25g Each for $378.66
Description & Specifications

Specifications

Formula Weight 257.09
Chemical Name or Material 3-(4-Bromobenzoyl)propionic acid
Assay ≥98%
Molecular Formula C10H9BrO3
MDL Number MFCD00016563
CAS 6340-79-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 145° to 148°C
Beilstein 2104410
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.