missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Bromobenzoyl)propionic acid, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L0400814

 View more versions of this product

Catalog No. AAL0400814



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-(4-Bromobenzoyl)propionic acid
145°C to 148°C
MFCD00016563
2104410
ZODFRCZNTXLDDW-UHFFFAOYSA-N
4-(4-bromophenyl)-4-oxobutanoic acid
80646
≥98%
6340-79-0
C10H9BrO3
25 g
3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
OC(=O)CCC(=O)C1=CC=C(Br)C=C1
257.08
257.09

Chemical Identifiers

6340-79-0
257.08
ZODFRCZNTXLDDW-UHFFFAOYSA-N
80646
OC(=O)CCC(=O)C1=CC=C(Br)C=C1
C10H9BrO3
MFCD00016563
3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
4-(4-bromophenyl)-4-oxobutanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-732-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only