missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-Bromobenzoyl)propionic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 6340-79-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9BrO3
Linear Formula BrC6H4CO(CH)COOH
Formula Weight 257.08
MDL Number MFCD00016563
Chemical Name or Material 3-(4-Bromobenzoyl)propionic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC344340250 SDS Fisher Scientific Acros
344340250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6340-79-0
100.0
C10H9BrO3
257.08
3-(4-Bromobenzoyl)propionic acid
96.0
97%
BrC6H4CO(CH)COOH
MFCD00016563
≥96.0%

Specifications

Shiny Crystalline Flakes or Powder
25g
100.0
BrC6H4CO(CH)COOH
6340-79-0
MFCD00016563
Glass bottle
≥96.0%
257.08
White to light beige
96.0
C10H9BrO3
97%
Authentic
3-(4-Bromobenzoyl)propionic acid

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.