Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Molecular Formula C22H35BN2O4
Formula Weight 402.34
MDL Number MFCD16294542
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5193903

 
alfa aesar™
H5193903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5193906

 
alfa aesar™
H5193906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 402.34
Chemical Name or Material 3-(4-Boc-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C22H35BN2O4
MDL Number MFCD16294542
Quantity 1g

Notes
Product of Advanced Molecular Technologies Pty Ltd.