Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 96%

$338.64 - $338.64

Chemical Identifiers

Chemical Name or Material 3-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Molecular Formula C22H33BN2O5
Formula Weight 416.32
MDL Number MFCD09266185
Synonym tert-Butyl 3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2927503

 
alfa aesar™
H2927503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2927506

 
alfa aesar™
H2927506
5g Each for $338.64
Description & Specifications

Specifications

Formula Weight 416.32
Chemical Name or Material 3-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Assay 96%
Molecular Formula C22H33BN2O5
MDL Number MFCD09266185
Synonym tert-Butyl 3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
Quantity 1g