missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4'-Bis(trifluoromethyl)benzophenone, 99%

Manufacturer:  Alfa Aesar™ L0964214

 View more versions of this product

Catalog No. AAL0964214



Specifications

Specifications

3,4'-Bis(trifluoromethyl)benzophenone
25g
C15H8F6O
3064970
HWBXNAPXNCBCQO-UHFFFAOYSA-N
[3-(trifluoromethyl)phenyl]-[4-(trifluoromethyl)phenyl]methanone
736046
99%
93°C to 95°C
21084-22-0
MFCD00013561
3,4'-bis trifluoromethyl benzophenone, 3,4'-di trifluoromethyl benzophenone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanone, 3-trifluoromethyl phenyl-4-trifluoromethyl phenyl methanone, methanone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl, 3,4-bis trifluoromethyl benzophenone, benzophenone, 3,4'-bis trifluoromethyl
C1=CC(=CC(=C1)C(F)(F)F)C(=O)C2=CC=C(C=C2)C(F)(F)F
318.218
318.22

Chemical Identifiers

21084-22-0
318.218
HWBXNAPXNCBCQO-UHFFFAOYSA-N
736046
C1=CC(=CC(=C1)C(F)(F)F)C(=O)C2=CC=C(C=C2)C(F)(F)F
C15H8F6O
MFCD00013561
3,4'-bis trifluoromethyl benzophenone, 3,4'-di trifluoromethyl benzophenone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanone, 3-trifluoromethyl phenyl-4-trifluoromethyl phenyl methanone, methanone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl, 3,4-bis trifluoromethyl benzophenone, benzophenone, 3,4'-bis trifluoromethyl
[3-(trifluoromethyl)phenyl]-[4-(trifluoromethyl)phenyl]methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents