missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenylyl)-N-Fmoc-L-alanine, 95%

CAS: 199110-64-0 | C30H25NO4 | 463.533 g/mol

Supplier:  Thermo Scientific Chemicals H5200203

 View more versions of this product

Catalog No. AAH5200203


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(4-Biphenylyl)-N-Fmoc-L-alanine
C30H25NO4
1 g
VSGACONKQRJFGX-NDEPHWFRSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
2761797
95%
199110-64-0
MFCD00191198
fmoc-l-4,4'-biphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-bip 4,4'-oh, fmoc-4-biphenyl-l-ala, fmoc-4-phenyl-phe-oh, 3-4-biphenylyl-n-fmoc-l-alanine, fmoc-p-phenyl-l-phenylalanine, fmoc-bip-oh, fmoc-l-phe 4-ph-oh, 2s-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
463.533
463.52

Chemical Identifiers

199110-64-0
463.533
VSGACONKQRJFGX-NDEPHWFRSA-N
2761797
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
C30H25NO4
MFCD00191198
fmoc-l-4,4'-biphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1,1'-biphenyl-4-yl propanoic acid, fmoc-bip 4,4'-oh, fmoc-4-biphenyl-l-ala, fmoc-4-phenyl-phe-oh, 3-4-biphenylyl-n-fmoc-l-alanine, fmoc-p-phenyl-l-phenylalanine, fmoc-bip-oh, fmoc-l-phe 4-ph-oh, 2s-3-1,1'-biphenyl-4-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylphenyl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only