missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Biphenylyl)-D-alanine, 98%

$140.03 - $419.46

Chemical Identifiers

CAS 170080-13-4
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD01318879
InChI Key JCZLABDVDPYLRZ-CQSZACIVSA-N
Synonym r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid
PubChem CID 7006711
IUPAC Name (2R)-2-amino-3-(4-phenylphenyl)propanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63785MD View Documents Alfa Aesar™
H63785MD
250mg Each for $140.03
 
AAH6378503 View Documents Alfa Aesar™
H6378503
1g Each for $419.46
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

170080-13-4
241.29
JCZLABDVDPYLRZ-CQSZACIVSA-N
7006711
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N
C15H15NO2
MFCD01318879
r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid
(2R)-2-amino-3-(4-phenylphenyl)propanoic acid

Specifications

170080-13-4
MFCD01318879
JCZLABDVDPYLRZ-CQSZACIVSA-N
(2R)-2-amino-3-(4-phenylphenyl)propanoic acid
7006711
Powder
White
3-(4-Biphenylyl)-D-alanine
C15H15NO2
r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N
241.29
241.29
98%
250mg