missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-(4-Biphenylyl)-D-alanine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6378503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
3-(4-Biphenylyl)-D-alanine | |
White | |
MFCD01318879 | |
Slightly soluble in water. | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N | |
241.29 | |
241.29 | |
1 g |
170080-13-4 | |
C15H15NO2 | |
r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid | |
JCZLABDVDPYLRZ-CQSZACIVSA-N | |
(2R)-2-amino-3-(4-phenylphenyl)propanoic acid | |
7006711 | |
98% | |
Powder |
Chemical Identifiers
170080-13-4 | |
241.29 | |
JCZLABDVDPYLRZ-CQSZACIVSA-N | |
7006711 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N |
C15H15NO2 | |
MFCD01318879 | |
r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid | |
(2R)-2-amino-3-(4-phenylphenyl)propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures