Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Aminophenyl)propionic acid, 97%

$78.80 - $305.88

Chemical Identifiers

Chemical Name or Material 3-(4-Aminophenyl)propionic acid
CAS 2393-17-1
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00017118
Beilstein 2209840
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1933006

  SDS
alfa aesar™
L1933006
5g Each for $305.88

AAL1933003

  SDS
alfa aesar™
L1933003
1g Each for $78.80
Description & Specifications

Specifications

Formula Weight 165.19
MDL Number MFCD00017118
Chemical Name or Material 3-(4-Aminophenyl)propionic acid
Assay 97%
CAS 2393-17-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
Molecular Formula C9H11NO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 133° to 135°C
Beilstein 2209840
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.