missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,9,10-Perylenetetracarboxylic diimide

Manufacturer:  Alfa Aesar™ 4409806

 View more versions of this product

Catalog No. AA4409806



Specifications

Specifications

3,4,9,10-Perylenetetracarboxylic diimide
81-33-4
MFCD00024144
Insoluble in water.
C1=CC2=C3C(=CC=C4C3=C1C5=C6C4=CC=C7C6=C(C=C5)C(=O)NC7=O)C(=O)NC2=O
66475
390.35
5g
C24H10N2O4
3,4,9,10-perylenetetracarboxylic acid diimide
KJOLVZJFMDVPGB-UHFFFAOYSA-N
390.354
CHEBI:52753
Powder

Chemical Identifiers

81-33-4
390.354
KJOLVZJFMDVPGB-UHFFFAOYSA-N
66475
C1=CC2=C3C(=CC=C4C3=C1C5=C6C4=CC=C7C6=C(C=C5)C(=O)NC7=O)C(=O)NC2=O
C24H10N2O4
MFCD00024144
3,4,9,10-perylenetetracarboxylic acid diimide
CHEBI:52753
Safety and Handling

Safety and Handling

EINECSNumber : 201-344-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents