Please login to your online account to display your discounted pricing

3,4,9,10-Perylenetetracarboxylic Dianhydride 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  130081000

 View more versions of this product

Catalog No. AC130081000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥97.5%
Formula Weight 392.32
Quantity 100g
Color Dark red
EINECS Number 204-905-3
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 1 MOISTURE SENSITIVE
Water 1% max. (K.F.)
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic dianhydride
Name Note 98%
Assay Percent Range 98%
Molecular Formula C24H8O6
MDL Number MFCD00006916
CAS 128-69-8

Specifications

Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic dianhydride
Name Note 98%
CAS 128-69-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C24H8O6
Formula Weight 392.32
MDL Number MFCD00006916
Accessories

MOISTURE SENSITIVE

SDS & Safety Documents