missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3(4),8(9)-Bis(aminomethyl)tricyclo[5.2.1.0(2,6)]decane, TCI America™

$53.47 - $328.48

Chemical Identifiers

CAS 76364-76-6
Molecular Formula C12H22N2
Molecular Weight (g/mol) 194.32
MDL Number MFCD00161489
InChI Key LQGTVLDXRIBHQO-UHFFFAOYNA-N
Synonym Octahydro-4,7-methanoindene-1(2),5(6)-dimethanamine
PubChem CID 101136807
SMILES CCN.CCN.C1CC2C3CCC(C3)C2C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B144725G View Documents TCI America
B144725G
25g Each for $53.47
 
B1447500G View Documents TCI America
B1447500G
500g Each for $328.48
 
Specifications

Chemical Identifiers

76364-76-6
194.32
LQGTVLDXRIBHQO-UHFFFAOYNA-N
101136807
C12H22N2
MFCD00161489
Octahydro-4,7-methanoindene-1(2),5(6)-dimethanamine
CCN.CCN.C1CC2C3CCC(C3)C2C1

Specifications

Yellow
76364-76-6
MFCD00161489
Octahydro-4,7-methanoindene-1(2),5(6)-dimethanamine
CCN.CCN.C1CC2C3CCC(C3)C2C1
101136807
Clear Liquid at 20°C
3(4),8(9)-Bis(aminomethyl)tricyclo[5.2.1.0(2,6)]decane
25g
C12H22N2
2735
LQGTVLDXRIBHQO-UHFFFAOYNA-N
194.32
194.32
≥97.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1493

TSCA : No