missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3',4',7,8-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L13829MD

 View more versions of this product

Catalog No. AAL13829MD


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3',4',7,8-Tetramethoxyflavone
206°C to 208°C
MFCD00143001
330821
ZRLWYUNKZNRQLO-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
4033898
97%
65548-55-2
C19H18O6
250 mg
3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC
342.347
342.35

Chemical Identifiers

65548-55-2
342.347
ZRLWYUNKZNRQLO-UHFFFAOYSA-N
4033898
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C=C3)OC)OC)OC
C19H18O6
MFCD00143001
3',4',7,8-tetramethoxyflavone, 2-3,4-dimethoxyphenyl-7,8-dimethoxy-4h-chromen-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxychromen-4-one, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-7,8-dimethoxy, 2-3,4-dimethoxy-phenyl-7,8-dimethoxy-chromen-4-one, acmc-1b8pz, 7,8,3',4'-tetramethoxyflavone, 7,8-dimethoxy-2-3,4-dimethoxyphenyl-4h-chromen-4-one
2-(3,4-dimethoxyphenyl)-7,8-dimethoxychromen-4-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only