Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4',7,8-Tetrahydroxyflavone hydrate, 97%

$116.77 - $349.57

Chemical Identifiers

Chemical Name or Material 3,4',7,8-Tetrahydroxyflavone hydrate
Molecular Formula C15H10O6·xH2O
Formula Weight 286.24 (Anhydrous)
MDL Number MFCD11044443
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2734803

  SDS
alfa aesar™
H2734803
1g Each for $116.77

AAH2734806

  SDS
alfa aesar™
H2734806
5g Each for $349.57
Description & Specifications

Specifications

Formula Weight 286.24 (Anhydrous)
MDL Number MFCD11044443
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C15H10O6·xH2O
Melting Point >300°C
Quantity 1g
Chemical Name or Material 3,4',7,8-Tetrahydroxyflavone hydrate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.