missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6027503
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesKeep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, water, moisture sensitive..
Specifications
3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal | |
0.928 | |
>113°C (235°F) | |
1.456 | |
1 g | |
tri-o-tert-butyldimethylsilyl-d-glucal, 3,4,6-tri-o-t-butyl-dimethylsilyl-d-glucal, 3-o,4-o,6-o-tris tert-butyldimethylsilyl-1,2-dideoxy-d-arabino-1-hexenopyranose, 2r,3r,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl bis oxy bis tert-butyldimethylsilane, tert-butyl 2r,3r,4r-4-tert-butyldimethylsilyl oxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl oxy dimethylsilane | |
−30° (c=1 in Chloroform) | |
[(2R,3R,4R)-3,4-bis[[tert-butyl(dimethyl)silyl]oxy]-3,4-dihydro-2H-pyran-2-yl]methoxy-tert-butyl-dimethylsilane | |
10648718 | |
97% |
79999-47-6 | |
210°C (1.2 mmHg) | |
C24H52O4Si3 | |
MFCD00171721 | |
Air and moisture sensitive | |
FMXIGEXRLSNRMR-NJYVYQBISA-N | |
CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C | |
488.931 | |
488.92 |
Chemical Identifiers
79999-47-6 | |
488.931 | |
FMXIGEXRLSNRMR-NJYVYQBISA-N | |
10648718 | |
CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C |
C24H52O4Si3 | |
MFCD00171721 | |
tri-o-tert-butyldimethylsilyl-d-glucal, 3,4,6-tri-o-t-butyl-dimethylsilyl-d-glucal, 3-o,4-o,6-o-tris tert-butyldimethylsilyl-1,2-dideoxy-d-arabino-1-hexenopyranose, 2r,3r,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl bis oxy bis tert-butyldimethylsilane, tert-butyl 2r,3r,4r-4-tert-butyldimethylsilyl oxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl oxy dimethylsilane | |
[(2R,3R,4R)-3,4-bis[[tert-butyl(dimethyl)silyl]oxy]-3,4-dihydro-2H-pyran-2-yl]methoxy-tert-butyl-dimethylsilane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen