missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4,6-Tri-O-methyl-D-glucal, 97%, Thermo Scientific Chemicals
$76.70 - $252.21
Chemical Identifiers
CAS | 16740-98-0 |
---|---|
Molecular Formula | C9H16O4 |
Molecular Weight (g/mol) | 188.22 |
MDL Number | MFCD22989010 |
InChI Key | PZJPNKKRMNFDQJ-UHFFFAOYNA-N |
Synonym | tri-o-methyl-d-glucal, 3,4,6-tri-o-methyl-d-glucal, tri-o-methyl glucal, 2r,3s,4r-3,4-dimethoxy-2-methoxymethyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-trimethyl-1,2-dideoxy-d-arabino-1-hexenopyranose |
PubChem CID | 11252452 |
SMILES | COCC1OC=CC(OC)C1OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH60694MD
|
Thermo Scientific Chemicals
H60694MD |
250 mg |
Each for $76.70
|
|
|||||
AAH6069403
|
Thermo Scientific Chemicals
H6069403 |
1 g |
Each for $252.21
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16740-98-0 | |
188.22 | |
PZJPNKKRMNFDQJ-UHFFFAOYNA-N | |
11252452 |
C9H16O4 | |
MFCD22989010 | |
tri-o-methyl-d-glucal, 3,4,6-tri-o-methyl-d-glucal, tri-o-methyl glucal, 2r,3s,4r-3,4-dimethoxy-2-methoxymethyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-trimethyl-1,2-dideoxy-d-arabino-1-hexenopyranose | |
COCC1OC=CC(OC)C1OC |
Specifications
16740-98-0 | |
MFCD22989010 | |
tri-o-methyl-d-glucal, 3,4,6-tri-o-methyl-d-glucal, tri-o-methyl glucal, 2r,3s,4r-3,4-dimethoxy-2-methoxymethyl-3,4-dihydro-2h-pyran, 3-o,4-o,6-o-trimethyl-1,2-dideoxy-d-arabino-1-hexenopyranose | |
COCC1OC=CC(OC)C1OC | |
188.22 | |
188.22 | |
3,4,6-Tri-O-methyl-D-glucal |
C9H16O4 | |
250 mg | |
PZJPNKKRMNFDQJ-UHFFFAOYNA-N | |
3,4-dimethoxy-2-(methoxymethyl)-3,4-dihydro-2H-pyran | |
11252452 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only