missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose, 97%

$55.51 - $175.80

Chemical Identifiers

CAS 132732-60-6
Molecular Formula C27H30O5
Molecular Weight (g/mol) 434.532
MDL Number MFCD06797170
InChI Key PDGHLARNGSMEJE-GWDBROLASA-N
Synonym 3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-ol, 2-deoxy-3,4,6-tri-o-benzyl-d-glucopyranose, 3-o,4-o,6-o-tribenzyl-2-deoxy-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, d-arabino-hexose,2-deoxy-3,4,6-tris-o-phenylmethyl, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol
PubChem CID 10972120
IUPAC Name (4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
SMILES C1C(C(C(OC1O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60180MD View Documents Alfa Aesar™
H60180MD
250mg Each for $55.51
 
AAH6018003 View Documents Alfa Aesar™
H6018003
1g Each for $175.80
 
Specifications

Chemical Identifiers

132732-60-6
434.532
PDGHLARNGSMEJE-GWDBROLASA-N
10972120
C1C(C(C(OC1O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
C27H30O5
MFCD06797170
3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-ol, 2-deoxy-3,4,6-tri-o-benzyl-d-glucopyranose, 3-o,4-o,6-o-tribenzyl-2-deoxy-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, d-arabino-hexose,2-deoxy-3,4,6-tris-o-phenylmethyl, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol
(4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol

Specifications

250mg
C27H30O5
3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-ol, 2-deoxy-3,4,6-tri-o-benzyl-d-glucopyranose, 3-o,4-o,6-o-tribenzyl-2-deoxy-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucopyranose, d-arabino-hexose,2-deoxy-3,4,6-tris-o-phenylmethyl, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl tetrahydro-2h-pyran-2-ol
C1C(C(C(OC1O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
434.532
434.52
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose
132732-60-6
MFCD06797170
PDGHLARNGSMEJE-GWDBROLASA-N
(4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-ol
10972120
97%
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C