missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,6-Tri-O-benzyl-2-deoxy-D-glucono-1,5-lactone, 97%

$60.15 - $189.68

Chemical Identifiers

CAS 72605-77-7
Molecular Formula C27H28O5
Molecular Weight (g/mol) 432.516
MDL Number MFCD22989012
InChI Key XDRGBAJBRHEMCV-SLQPCKNISA-N
Synonym 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-1-oxo-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-one, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one
PubChem CID 11048306
IUPAC Name (4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-one
SMILES C1C(C(C(OC1=O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60716MD View Documents Alfa Aesar™
H60716MD
250mg Each for $60.15
 
AAH6071603 View Documents Alfa Aesar™
H6071603
1g Each for $189.68
 
Specifications

Chemical Identifiers

72605-77-7
432.516
XDRGBAJBRHEMCV-SLQPCKNISA-N
11048306
C1C(C(C(OC1=O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
C27H28O5
MFCD22989012
4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-1-oxo-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-one, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one
(4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-one

Specifications

83°C to 88°C
72605-77-7
MFCD22989012
XDRGBAJBRHEMCV-SLQPCKNISA-N
(4R,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)oxan-2-one
11048306
97%
250mg
C27H28O5
4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 3-o,4-o,6-o-tribenzyl-1,2-dideoxy-1-oxo-d-glucopyranose, 3,4,6-tri-o-benzyl-2-deoxy-d-glucono-1,5-lactone, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxy methyl oxan-2-one, 4r,5s,6r-4,5-bis benzyloxy-6-benzyloxymethyl tetrahydro-2h-pyran-2-one
C1C(C(C(OC1=O)COCC2=CC=CC=C2)OCC3=CC=CC=C3)OCC4=CC=CC=C4
432.516
432.51
3,4,6-Tri-O-benzyl-2-deoxy-D-glucono-1,5-lactone
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C