missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,6-Tri-O-acetyl-D-galactal

Manufacturer:  Alfa Aesar™ J6690406

 View more versions of this product

Catalog No. AAJ6690406



Description

Description

Applications
3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

Solubility
Soluble in chloroform at 10mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3,4,6-Tri-O-acetyl-D-galactal
Yellow
5g
C12H16O7
1,3,4-tri-o-acetyl-2,6-anhydro-5-deoxyhex-5-enitol, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol triacetate, 2r,3r,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 2r,3r,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, 3,4,6-tri-o-acetyl-d-galactal, d-arabino-hex-5-enitol, 2,6-anhydro-5-deoxy-, triacetate, d-galactal triacetate, galactal triacetate, tri-o-acetyl-d-galactal, tri-o-acetylgalactal
LLPWGHLVUPBSLP-IJLUTSLNSA-N
[(2R,3R,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
640125
Oil or Solid
>110°C (235°F)
34°C to 38°C
4098-06-0
MFCD00064092
Soluble in chloroform at 10mg/ml
CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C
272.253
272.25

Chemical Identifiers

4098-06-0
272.253
LLPWGHLVUPBSLP-IJLUTSLNSA-N
640125
CC(=O)OCC1C(C(C=CO1)OC(=O)C)OC(=O)C
C12H16O7
MFCD00064092
1,3,4-tri-o-acetyl-2,6-anhydro-5-deoxyhex-5-enitol, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol triacetate, 2r,3r,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, 2r,3r,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, 3,4,6-tri-o-acetyl-d-galactal, d-arabino-hex-5-enitol, 2,6-anhydro-5-deoxy-, triacetate, d-galactal triacetate, galactal triacetate, tri-o-acetyl-d-galactal, tri-o-acetylgalactal
[(2R,3R,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : 223-859-5

TSCA : No

Recommended Storage : Keep cold

Documents