missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Tris(benzyloxy)benzoic Acid, TCI America™

Manufacturer:  TCI America T186925G

 View more versions of this product

Catalog No. T186925G



Specifications

Specifications

3,4,5-Tris(benzyloxy)benzoic Acid
196°C
1486-48-2
MFCD00825832
NRCGMEJBEDWPMG-UHFFFAOYSA-N
3,4,5-tris(phenylmethoxy)benzoic acid
11133969
Crystal-Powder at 20°C
White
25g
C28H24O5
3,4,5-Tribenzyloxybenzoic Acid
C1=CC=C(C=C1)COC2=CC(=CC(=C2OCC3=CC=CC=C3)OCC4=CC=CC=C4)C(=O)O
440.495
440.50
≥98.0% (HPLC,T)

Chemical Identifiers

1486-48-2
440.495
NRCGMEJBEDWPMG-UHFFFAOYSA-N
11133969
C1=CC=C(C=C1)COC2=CC(=CC(=C2OCC3=CC=CC=C3)OCC4=CC=CC=C4)C(=O)O
C28H24O5
MFCD00825832
3,4,5-Tribenzyloxybenzoic Acid
3,4,5-tris(phenylmethoxy)benzoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents