missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxytoluene, 98%

Manufacturer:  Alfa Aesar™ A1520122

 View more versions of this product

Catalog No. AAA1520122



Specifications

Specifications

3,4,5-Trimethoxytoluene
115°C to 116°C (4mmHg)
100g
C10H14O3
1953791
KCIZTNZGSBSSRM-UHFFFAOYSA-N
1,2,3-trimethoxy-5-methylbenzene
80922
182.22
74°C (165°F)
34°C to 38°C
6443-69-2
MFCD00008397
3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
CC1=CC(=C(C(=C1)OC)OC)OC
182.219
CHEBI:81354
98%

Chemical Identifiers

6443-69-2
182.219
KCIZTNZGSBSSRM-UHFFFAOYSA-N
80922
1,2,3-trimethoxy-5-methylbenzene
C10H14O3
MFCD00008397
3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
CHEBI:81354
CC1=CC(=C(C(=C1)OC)OC)OC
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents