Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxyphenylglyoxal hydrate, 98%, dry wt. basis

$524.35 - $524.35

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxyphenylglyoxal hydrate
CAS 150114-69-5
Molecular Formula C11H12O5·xH2O
Formula Weight 224.22 (Anhydrous)
MDL Number MFCD05664097
Beilstein 3733165
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2027006

  SDS
alfa aesar™
L2027006
5g Each for $524.35
Description & Specifications

Specifications

Formula Weight 224.22 (Anhydrous)
Chemical Name or Material 3,4,5-Trimethoxyphenylglyoxal hydrate
Assay 98%
Molecular Formula C11H12O5·xH2O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Notes (dry wt.)
MDL Number MFCD05664097
CAS 150114-69-5
Beilstein 3733165
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 126° to 139°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.