Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxyphenylacetone, 98%

$551.47 - $551.47

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxyphenylacetone
CAS 16603-18-2
Molecular Formula C12H16O4
Formula Weight 224.26
MDL Number MFCD00210454
Synonym 3,4,5-Trimethoxyphenyl-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1900614

  SDS
alfa aesar™
A1900614
25g Each for $551.47
Description & Specifications

Specifications

Formula Weight 224.26
Quantity 25g
Synonym 3,4,5-Trimethoxyphenyl-2-propanone
Melting Point 55° to 65°C
Assay 98%
Chemical Name or Material 3,4,5-Trimethoxyphenylacetone
Molecular Formula C12H16O4
MDL Number MFCD00210454
CAS 16603-18-2