Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxyphenyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxyphenyl isothiocyanate
CAS 35967-24-9
Molecular Formula C10H11NO3S
Formula Weight 225.27g/mol
MDL Number MFCD00041076
UN Number UN2923
Beilstein 2736554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1070903

 
alfa aesar™
L10709-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1070906

 
alfa aesar™
L10709-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 63° to 67°C
Quantity 1g
Chemical Name or Material 3,4,5-Trimethoxyphenyl isothiocyanate
CAS 35967-24-9
Assay 98%
Molecular Formula C10H11NO3S
Formula Weight 225.27g/mol
MDL Number MFCD00041076
UN Number UN2923
Beilstein 2736554
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.