Please login to your online account to display your discounted pricing

3,4,5-Trimethoxyphenol 98.5+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  189140050

 View more versions of this product

Catalog No. AC189140050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.50% (GC)
Formula Weight 184.19
Physical Form Powder and/or Chunks
Melting Point 143° to 149°C
Quantity 5g
Color Light yellow to beige to light brown
EINECS Number 211-387-2
TSCA TSCA
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Chemical Name or Material 3, 4, 5-Trimethoxyphenol
Molecular Formula C9H12O4
CAS 642-71-7
MDL Number MFCD00008389
Assay Percent Range 98.5+%
Name Note 98.5+%
Linear Formula (CH3O)3C6H2OH

Specifications

Chemical Name or Material 3, 4, 5-Trimethoxyphenol
Name Note 98.5+%
CAS 642-71-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.50% (GC)
Assay Percent Range 98.5+%
Molecular Formula C9H12O4
Linear Formula (CH3O)3C6H2OH
Formula Weight 184.19
MDL Number MFCD00008389
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents