Please login to your online account to display your discounted pricing

3,4,5-Trimethoxyphenol 98.5+%, ACROS Organics™

$118.85 - $118.85

Chemical Identifiers

Chemical Name or Material 3, 4, 5-Trimethoxyphenol
Name Note 98.5+%
CAS 642-71-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.50% (GC)
Assay Percent Range 98.5+%
Molecular Formula C9H12O4
Linear Formula (CH3O)3C6H2OH
Formula Weight 184.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189140250

  SDS
fisher scientific acros
189140250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC189140050

  SDS
thermo fisher scientific
189140050
Glass bottle 5g Each for $118.85

AC189141000

  SDS
thermo fisher scientific
AC189141000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.50% (GC)
Physical Form Powder and/or Chunks
Formula Weight 184.19
Melting Point 143° to 149°C
Quantity 25g
Color Light yellow to beige to light brown
EINECS Number 211-387-2
TSCA TSCA
CAS Max % 100.0
Packaging Glass bottle
Name Note 98.5+%
Assay Percent Range 98.5+%
Molecular Formula C9H12O4
MDL Number MFCD00008389
CAS 642-71-7
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 3, 4, 5-Trimethoxyphenol
Linear Formula (CH3O)3C6H2OH
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.