missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trimethoxybenzylamine, TCI America™

Manufacturer:  TCI America T308725G

 View more versions of this product

Catalog No. T308725G



Specifications

Specifications

3,4,5-Trimethoxybenzylamine
Yellow
18638-99-8
MFCD00017150
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
COC1=CC(=CC(=C1OC)OC)CN
197.234
197.23
≥97.0% (GC,T)
121°C
25g
C10H15NO3
2735
YUPUSBMJCFBHAP-UHFFFAOYSA-N
(3,4,5-trimethoxyphenyl)methanamine
87736
Clear Liquid at 20°C

Chemical Identifiers

18638-99-8
197.234
YUPUSBMJCFBHAP-UHFFFAOYSA-N
87736
COC1=CC(=CC(=C1OC)OC)CN
C10H15NO3
MFCD00017150
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
(3,4,5-trimethoxyphenyl)methanamine
Safety and Handling

Safety and Handling

TSCA : No

Documents