Please login to your online account to display your discounted pricing

3,4,5-Trimethoxybenzylamine, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35117-0250

 View more versions of this product

Catalog No. AC351170250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 197.24
Quantity 25g
Color Clear colorless to slightly yellow
Density 1.155g/mL
EINECS Number 242-468-0
TSCA TSCA
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 3, 4, 5-Trimethoxybenzylamine
Packaging Glass bottle
Refractive Index 1.5455 to 1.5485 (20°C, 589nm)
Name Note 96%
Assay Percent Range 96%
Molecular Formula C10H15NO3
Linear Formula (CH3O)3C6HCHNH
MDL Number MFCD00017150
CAS 18638-99-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3, 4, 5-Trimethoxybenzylamine
Name Note 96%
CAS 18638-99-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C10H15NO3
Linear Formula (CH3O)3C6HCHNH
Formula Weight 197.24
MDL Number MFCD00017150
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents