missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trimethoxybenzonitrile, TCI America™

Manufacturer:  TCI America T32445G

 View more versions of this product

Catalog No. T32445G



Specifications

Specifications

3,4,5-Trimethoxybenzonitrile
White-Yellow
5g
C10H11NO3
3,4,5-trimethoxybenzenecarbonitrile, benzonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxy-benzonitrile, pubchem13110, 3,4,5-trimethoxybenz, timtec-bb sbb007562, acmc-1bs5l, 2,5-trimethoxybenzonitrile, 3,5-trimethoxybenzonitrile, benzonitrile,4,5-trimethoxy
COC1=CC(=CC(=C1OC)OC)C#N
193.202
193.20
≥98.0% (GC)
185°C
94°C
1885-35-4
MFCD00001803
OSBQUSPVORCDCU-UHFFFAOYSA-N
3,4,5-trimethoxybenzonitrile
15892
Crystal-Powder at 20°C

Chemical Identifiers

1885-35-4
193.202
OSBQUSPVORCDCU-UHFFFAOYSA-N
15892
COC1=CC(=CC(=C1OC)OC)C#N
C10H11NO3
MFCD00001803
3,4,5-trimethoxybenzenecarbonitrile, benzonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxy-benzonitrile, pubchem13110, 3,4,5-trimethoxybenz, timtec-bb sbb007562, acmc-1bs5l, 2,5-trimethoxybenzonitrile, 3,5-trimethoxybenzonitrile, benzonitrile,4,5-trimethoxy
3,4,5-trimethoxybenzonitrile
Safety and Handling

Safety and Handling

RTECSNumber : DI4965000

TSCA : No

Documents