missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxybenzonitrile, 97%

Manufacturer:  Alfa Aesar™ A1463418

 View more versions of this product

Catalog No. AAA1463418



Specifications

Specifications

3,4,5-Trimethoxybenzonitrile
180°C to 185°C (10mmHg)
50g
C10H11NO3
979686
OSBQUSPVORCDCU-UHFFFAOYSA-N
3,4,5-trimethoxybenzonitrile
15892
97%
Characteristic
91°C to 95°C
1885-35-4
MFCD00001803
3,4,5-trimethoxybenzenecarbonitrile, benzonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxy-benzonitrile, pubchem13110, 3,4,5-trimethoxybenz, timtec-bb sbb007562, acmc-1bs5l, 2,5-trimethoxybenzonitrile, 3,5-trimethoxybenzonitrile, benzonitrile,4,5-trimethoxy
COC1=CC(=CC(=C1OC)OC)C#N
193.202
193.2

Chemical Identifiers

1885-35-4
193.202
OSBQUSPVORCDCU-UHFFFAOYSA-N
15892
COC1=CC(=CC(=C1OC)OC)C#N
C10H11NO3
MFCD00001803
3,4,5-trimethoxybenzenecarbonitrile, benzonitrile, 3,4,5-trimethoxy, 3,4,5-trimethoxy-benzonitrile, pubchem13110, 3,4,5-trimethoxybenz, timtec-bb sbb007562, acmc-1bs5l, 2,5-trimethoxybenzonitrile, 3,5-trimethoxybenzonitrile, benzonitrile,4,5-trimethoxy
3,4,5-trimethoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning

RTECSNumber : DI4965000

Documents