Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxybenzonitrile, 97%

$219.99 - $219.99

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxybenzonitrile
CAS 1885-35-4
Molecular Formula C10H11NO3
Formula Weight 193.2
MDL Number MFCD00001803
Beilstein 979686
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1463418

  SDS
alfa aesar™
A1463418
50g Each for $219.99

AAA1463409

  SDS
alfa aesar™
A1463409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,4,5-Trimethoxybenzonitrile
Odor Characteristic
Boiling Point 180° to 185°C (10mmHg)
Melting Point 91° to 95°C
Quantity 50g
CAS 1885-35-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
RTECS Number DI4965000
Assay 97%
Molecular Formula C10H11NO3
Formula Weight 193.2
MDL Number MFCD00001803
Beilstein 979686
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.