Learn More
3,4,5-Trimethoxybenzoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139931000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 4, 5-Trimethoxybenzoic acid | |
118-41-2 | |
100 | |
Colorless to Yellow | |
Authentic | |
Plastic bottle | |
(CH3O)3C6H2CO2H | |
100 g | |
eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f | |
SJSOFNCYXJUNBT-UHFFFAOYSA-N | |
3,4,5-trimethoxybenzoic acid | |
8357 | |
212.2 | |
Liquid |
0.99 | |
98.5 | |
168.0°C to 172.0°C | |
225.0°C to 227.0°C (10.0 mmHg) | |
98.5% min. (HPLC) | |
C10H12O5 | |
MFCD00002501 | |
10, 481 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ethylether,chloroform | |
COC1=CC(=CC(OC)=C1OC)C(O)=O | |
212.2 | |
CHEBI:454991 | |
0.99 |
Chemical Identifiers
118-41-2 | |
212.2 | |
SJSOFNCYXJUNBT-UHFFFAOYSA-N | |
8357 | |
3,4,5-trimethoxybenzoic acid |
C10H12O5 | |
MFCD00002501 | |
eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f | |
CHEBI:454991 | |
COC1=CC(=CC(OC)=C1OC)C(O)=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 204-248-2
TSCA : TSCA
RUO – Research Use Only