Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxybenzhydrazide, 98%

$72.93 - $72.93

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxybenzhydrazide
CAS 3291-03-0
Molecular Formula C10H14N2O4
Formula Weight 226.23
MDL Number MFCD00014758
Beilstein 484531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2005506

  SDS
alfa aesar™
B2005506
5g Each for $72.93
Description & Specifications

Specifications

Formula Weight 226.23
CAS 3291-03-0
Molecular Formula C10H14N2O4
Chemical Name or Material 3,4,5-Trimethoxybenzhydrazide
Beilstein 484531
Melting Point 158° to 160°C
Quantity 5g
Assay 98%
MDL Number MFCD00014758