Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trimethoxybenzamide, 98%

$67.88 - $198.57

Chemical Identifiers

Chemical Name or Material 3,4,5-Trimethoxybenzamide
CAS 3086-62-2
Molecular Formula C10H13NO4
Formula Weight 211.22
MDL Number MFCD00017129
Beilstein 2697325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1215514

  SDS
alfa aesar™
L1215514
25g Each for $67.88

AAL1215522

  SDS
alfa aesar™
L1215522
100g Each for $198.57
Description & Specifications

Specifications

Formula Weight 211.22
Chemical Name or Material 3,4,5-Trimethoxybenzamide
Assay 98%
Molecular Formula C10H13NO4
MDL Number MFCD00017129
CAS 3086-62-2
Beilstein 2697325
Quantity 25g
TSCA TSCA
RTECS Number CV6125000
Melting Point 178° to 180°C