missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4,5-Trimethoxyaniline 98.0+%, TCI America™
$112.81 - $527.56
Chemical Identifiers
CAS | 24313-88-0 |
---|---|
Molecular Formula | C9H13NO3 |
Molecular Weight (g/mol) | 183.207 |
MDL Number | MFCD00008393 |
InChI Key | XEFRNCLPPFDWAC-UHFFFAOYSA-N |
Synonym | benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline |
PubChem CID | 32285 |
IUPAC Name | 3,4,5-trimethoxyaniline |
SMILES | COC1=CC(=CC(=C1OC)OC)N |
Chemical Identifiers
24313-88-0 | |
183.207 | |
XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
32285 | |
COC1=CC(=CC(=C1OC)OC)N |
C9H13NO3 | |
MFCD00008393 | |
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
3,4,5-trimethoxyaniline |
Specifications
24313-88-0 | |
C9H13NO3 | |
5 g | |
XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
3,4,5-trimethoxyaniline | |
32285 | |
≥98.0% (GC,T) | |
3,4,5-Trimethoxyaniline |
112°C | |
MFCD00008393 | |
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
COC1=CC(=CC(=C1OC)OC)N | |
183.207 | |
183.21 | |
Crystalline Powder |
Safety and Handling
TSCA : Yes