missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyaniline, 98+%

CAS: 24313-88-0 | C9H13NO3 | 183.207 g/mol

$92.68 - $257.74

Chemical Identifiers

CAS 24313-88-0
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00008393
InChI Key XEFRNCLPPFDWAC-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
PubChem CID 32285
IUPAC Name 3,4,5-trimethoxyaniline
SMILES COC1=CC(=CC(=C1OC)OC)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0161906
View Documents
Thermo Scientific Chemicals
L0161906
5 g N/A
Only null left
 
AAL0161914
View Documents
Thermo Scientific Chemicals
L0161914
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24313-88-0
183.207
XEFRNCLPPFDWAC-UHFFFAOYSA-N
32285
COC1=CC(=CC(=C1OC)OC)N
C9H13NO3
MFCD00008393
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
3,4,5-trimethoxyaniline

Specifications

24313-88-0
C9H13NO3
5 g
642919
XEFRNCLPPFDWAC-UHFFFAOYSA-N
3,4,5-trimethoxyaniline
32285
≥98%
110°C to 113°C
MFCD00008393
UN2811
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
COC1=CC(=CC(=C1OC)OC)N
183.207
183.21
3,4,5-Trimethoxyaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 246-154-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only